4-Nitrophenethyl bromide ≥98.0%

Supplier: TCI America

N0591-100G
76220-354EA 415.54 CAD
76220-354
4-Nitrophenethyl bromide ≥98.0%
4-Nitrophenethyl bromide

CAS Number: 5339-26-4
MDL Number: MFCD00007386
Molecular Formula: C8H8BrNO2
Molecular Weight: 230.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 69


Other Product Names / Synonyms: 4-Nitrophenethyl Bromide

Formula: C₈H₈BrNO₂
Melting Pt: 69 °C
MDL Number: MFCD00007386
CAS Number: 5339-26-4
EINECS: 226-271-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d39e6ea864d9b9585175a5d39f4b686140c2aba&vid=7d39e6ea864d9b9585175a5d39f4b686140c2aba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR