1-Methyl-3-octylimidazolium bis(trifluoromethylsulfonyl)imide ≥97.0% (by HPLC)

Supplier: TCI America
Warning

M3039-25G M3039-5G
76220-338EA 475.58 CAD
76220-338 76220-340
1-Methyl-3-octylimidazolium bis(trifluoromethylsulfonyl)imide ≥97.0% (by HPLC)
1-Methyl-3-octylimidazolium bis(trifluoromethylsulphonyl)imide
Formula: C₁₄H₂₃F₆N₃O₄S₂
MDL Number: MFCD08458946
CAS Number: 178631-04-4

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (HPLC) min. 97.0 area%
Purity (Ion exchange titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR