Boc-Asp-OMe ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

M2970-5G
76220-316EA 91.64 CAD
76220-316
Boc-Asp-OMe ≥97.0% (by HPLC, titration analysis)
Boc-Asp-OMe
Formula: C₁₀H₁₇NO₆
MDL Number: MFCD03094777
CAS Number: 98045-03-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 87.0 to 91.0 deg-C
Specific rotation [a]20/D -20.0 to -23.0 deg (C=1, MeOH)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR