1-Methylpyrrole-2-carbonitrile ≥98.0% (by GC)

Supplier: TCI America
Warning

M2784-1G M2784-5G
76220-258EA 180.12 CAD
76220-258 76220-260
1-Methylpyrrole-2-carbonitrile ≥98.0% (by GC)
1-Methylpyrrole-2-carbonitrile
Formula: C₆H₆N₂
Boiling Pt: 87°C
MDL Number: MFCD02693721
CAS Number: 34884-10-1

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR