1-Methyl-5-(trifluoromethyl)pyrazole-4-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M2753-1G M2753-200MG
76220-244EA 349.18 CAD
76220-244 76220-246
1-Methyl-5-(trifluoromethyl)pyrazole-4-carboxylic acid ≥98.0% (by GC, titration analysis)
1-Methyl-5-(trifluoromethyl)pyrazole-4-carboxylic acid
Formula: C₆H₅F₃N₂O₂
MDL Number: MFCD04110182
CAS Number: 119083-00-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 121.0 to 125.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR