(5-Fluoro-2-nitrophenyl)(2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate ≥95.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: (5-Fluoro-2-nitrophenyl)(2,4,6-trimethoxyphenyl)iodonium Tosylate

F1110-200MG
76220-094EA 188.02 CAD
76220-094
(5-Fluoro-2-nitrophenyl)(2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate ≥95.0% (by HPLC)
(5-Fluoro-2-nitrophenyl)(2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate
Formula: C₂₂H₂₁FINO₈S
MW: 605.37 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30475654
CAS Number: 1868173-33-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 95.0 % (total of Ion-Pair)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR