1-Ethyl-3-methylpyridinium bis(trifluoromethanesulfonyl)imide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

E0756-5G
76220-020EA 164.32 CAD
76220-020
1-Ethyl-3-methylpyridinium bis(trifluoromethanesulfonyl)imide ≥98.0% (by HPLC, titration analysis)
1-Ethyl-3-methylpyridinium bis(trifluoromethanesulfonyl)imide
Formula: C₁₀H₁₂F₆N₂O₄S₂
Storage Temperature: Refrigerator
CAS Number: 841251-37-4

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green clear liquid
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR