1,4-Di(1-pyrenyl)benzene ≥98.0% (by HPLC)

Supplier: TCI America

D4922-1G D4922-200MG
76219-754EA 679.4 CAD
76219-754 76219-756
1,4-Di(1-pyrenyl)benzene ≥98.0% (by HPLC)
1,4-Di(1-pyrenyl)benzene
Formula: C₃₈H₂₂
MDL Number: MFCD12022457
CAS Number: 475460-77-6

Order Now

Specification Test Results

Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR