1,4-Difluoro-2-methyl-5-nitrobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

D4806-1G D4806-5G
76219-704EA 109.02 CAD
76219-704 76219-706
1,4-Difluoro-2-methyl-5-nitrobenzene ≥98.0% (by GC)
1,4-Difluoro-2-methyl-5-nitrobenzene
Formula: C₇H₅F₂NO₂
MDL Number: MFCD06200905
CAS Number: 141412-60-4

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR