1,6-Bis(acryloyloxy)-2,2,3,3,4,4,5,5-octafluorohexane ≥90.0% (by GC) stabilized

Supplier: TCI America


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

B5278-1G B5278-5G
76219-556EA 131.14 CAD
76219-556 76219-558
1,6-Bis(acryloyloxy)-2,2,3,3,4,4,5,5-octafluorohexane ≥90.0% (by GC) stabilized
1,6-Bis(acryloyloxy)-2,2,3,3,4,4,5,5-octafluorohexane
Formula: C₁₂H₁₀F₈O₄
MDL Number: MFCD00078412
CAS Number: 2264-01-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR