1,4-Bis(dimethylsilyl)benzene ≥98.0% (by GC)

Supplier: TCI America
Warning

B4748-25ML B4748-5ML
76219-426EA 341.28 CAD
76219-426 76219-428
1,4-Bis(dimethylsilyl)benzene ≥98.0% (by GC)
1,4-Bis(dimethylsilyl)benzene
Formula: C₁₀H₁₈Si₂
Boiling Pt: 220°C
Flash Pt: 85°C
MDL Number: MFCD00039790
CAS Number: 2488-01-9
EINECS: 219-638-8

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR