2,6-Dihydroxyanthracene ≥95.0% (by GC)

Supplier: TCI America

D5151-1G D5151-200MG
76219-188EA 537.2 CAD
76219-188 76219-190
2,6-Dihydroxyanthracene ≥95.0% (by GC)
2,6-Dihydroxyanthracene
Formula: C₁₄H₁₀O₂
Melting Pt: 300°C
MDL Number: MFCD24713437
CAS Number: 101488-73-7

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR