Distilling Head, Ace Glass
Supplier: Ace Glass
6608-10
76214-410EA 664.15 CAD
76214-410
Distilling Head, Ace Glass
Distillation Components Distillation Heads
Distilling head sits on top of your vapor path column. Has condenser side arm with drip tip.
  • High-quality borosilicate glass
  • 24/40 standard taper bottom joint and side arm joint
  • #11 Ace-Thred inlet/outlet for use with Ace-Safe hose connections
Can be joined with Ace short stem vacuum take-off adapters.