KIMBLE® SPE Disk Glassware and Manifolds, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
971010-3047
76213-994CS 6733.24 CAD
76213-994
KIMBLE® SPE Disk Glassware and Manifolds, DWK Life Sciences
Manifolds
The SPE glassware and manifolds have been specially designed for use with solid phase extraction disks to give you the highest possible recoveries of extracted analytes from drinking water, surface water and waste water samples.
  • Valves have stainless steel bodies with PTFE plugs
  • Extracted samples can be collected directly into K-D concentrator tubes
  • EPA water analysis vials, sample vials and test tubes up to 30 mm OD×100 mm high
  • PTFE-coated stainless steel support screen reduces the possibility of sample-to-sample cross-contamination compared to the
More Product Information