Savant™ SpeedVac™ Medium Capacity Vacuum Concentrators, Thermo Scientific
Supplier: Thermo Fisher Scientific
SPD140DDA-115 SPD140DDA-230 SPD140P1-230 SPD140P2-230
76210-426EA 26228 CAD
76210-426 76210-428 76210-572 76210-576
Savant™ SpeedVac™ Medium Capacity Vacuum Concentrators, Thermo Scientific
Concentrators
Savant™ SpeedVac™ medium capacity vacuum concentrators evaporate strong acids, bases, and aggressive organic solvents. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.
  • Equipped with radiant lamps within the chamber
  • Heated lid for processing high boiling points (DMSO, DMF) and independent timers for both heat and run
  • Audible alarm to alert user when operation is finished
  • Built-in vacuum gauge and vacuum display
  • Wider vapor port and tubing for enhanced vapor throughput,
More Product Information