TSX Series High-Performance Blood Bank Refrigerators, Automatic Defrost, Thermo Scientific
Supplier: Thermo Fisher Scientific
TSX2304BA TSX2304BD TSX5004BA TSX5004BD TSX1204BA TSX1204BD TSX3004BD TSX3004BA
75839-040EA 25280 CAD
75839-040 75839-092 75839-100 76208-598 76208-600 76320-226 76320-228 76320-230 76320-232 76320-308 76320-316
TSX Series High-Performance Blood Bank Refrigerators, Automatic Defrost, Thermo Scientific
Refrigerators
Thermo Scientific™ TSX Series high-performance blood bank refrigerators offer a sustainable choice for whole blood and blood components refrigeration.
  • Stores up to 385 blood bags (450 ml whole blood)
  • Whisper quiet with a sound level of 52 dBA
  • V-drive technology and Smart View compatibility
  • Up to 40% less energy usage than conventional refrigerant
  • Convenient, fully extendable and adjustable stainless steel drawers
  • Compatible with wireless monitoring systems and remote alarm contacts
  • Standard door locks provide comfort and assurance
  • Ergonomic door handle
More Product Information