Hercules® 400R6E Gloves, HexArmor
Supplier: HexArmor
400R6E-XL(10) 400R6E-XXL(11) 400R6E-M(8) 400R6E-L(9)
76208-030PR 825.16 CAD
76208-030 76208-032 76208-470 76208-472
Hercules® 400R6E Gloves, HexArmor
Gloves
The Hercules® 400R6E are no stranger to dangerous applications such as handling razor wire or protecting from animal bites. Gauntlet design and pre-curved shape for maximum comfort and protection.
  • SuperFabric®* brand material palm provides ANSI/ISEA cut level A9 resistance for some of the highest cut protection available in the industry (over entire hand)
  • Innovative industrial puncture protection over the entire hand from wires, metal, wood, glass, and other puncture related hazards
  • Gauntlet design and pre-curved shape for maximum comfort and dexterity
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Ambidextrous/Hand-specific Hand-specific
Disposable / reusable Reusable
Latex/Latex-free Latex-free
Powdered/Powder-free Powder-free
EN norm En388 4534