Chrome Series® 4027 Gloves, HexArmor
Supplier: HexArmor
4027-S(7) 4027-M(8) 4027-L(9) 4027-XL(10) 4027-XXL(11)
76207-930PR 162.61 CAD
76207-930 76207-932 76207-934 76207-936 76207-938 76207-940
Chrome Series® 4027 Gloves, HexArmor
Gloves
The Chrome Series® 4027 features a SuperFabric® brand material palm that provides ANSI/ISEA level A8 cut resistance giving over 5000 grams of protection, 20 times the amount offered by a standard leather glove.
  • SuperFabric®* brand material palm provides ANSI/ISEA level A8 cut resistance for some of the highest cut-resistance available in the industry
  • Superior back of hand impact protection system utilizes an advanced design to dissipate forceful blows over a large area
  • Durable TP-X® palm utilizes the highest-level abrasion resistance while maintaining an
More Product Information