Rig Lizard® 2030 Gloves, HexArmor
Supplier: HexArmor
2030X-S(7)
76207-892PR 161.82 CAD
76207-892 76207-900 76207-902
Rig Lizard® 2030 Gloves, HexArmor
Gloves
The Rig Lizard® 2030 delivers maximum grip and dexterity without sacrificing protection. It features a hi-vis silicone palm for superior grip and heat resistance, and the patented high-flex IR-X® Impact Exoskeleton™ protects the entire back- of-hand from smash and pinch injuries.
  • SuperFabric® brand material palm provides ANSI/ISEA cut level A6 resistance for some of the highest cut protection available in the industry (interior layer)
  • Glove was tested to the EN407 standard and received a level 3
  • Durable synthetic leather palm with heat-resistant silicone
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Ambidextrous/Hand-specific Hand-specific
Disposable / reusable Reusable
Latex/Latex-free Latex-free
Powdered/Powder-free Powder-free
EN norm En388 4532 En388 4532