VS350 Protective Eyewear, HexArmor
Supplier: HexArmor
12-11002-04
76207-846EA 76.16 CAD
76207-846 76207-848 76318-180
VS350 Protective Eyewear, HexArmor
Safety Glasses
Safety eyewear should never be treated as one size fits all. Simply wearing any generic pair of safety eyewear isn’t enough to protect the eyes.
  • Compact design for compatibility with other PPE
  • Low-profile, wraparound adaptive design fits a wide range of facial shapes
  • Wide adjustable head strap for secure, pressure-free fit
  • Indirect ventilation system
  • LTX™ Lens Technology for premium optical clarity
  • 100% UV protection
  • D3/D4 Liquid splash rated
The VS350 with permanent anti-fog coatings. This wide-vision, compact safety goggle offers ultimateMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable