Vactrap™ 2 Vacuum Trap System for Aspiration and Vacuum Protection, Foxx Life Sciences
Supplier: Foxx Life Sciences
302-2001-FLS 302-2101-FLS 302-4001-FLS 305-2001-FLS 305-2101-FLS 305-4001-FLS 302-2002-FLS 302-4002-FLS 305-2002-FLS 305-4002-FLS 300-0001-FLS 300-0002-FLS 300-0003-FLS 300-0005-FLS 25T-3418-FLS
76207-600EA 523.06 CAD
76207-600 76207-602 76207-604 76207-618 76207-620 76207-622 76207-624 76207-626 76207-628 76207-630 76207-632 76207-634 76207-636 76207-638 76207-640
Vactrap™ 2 Vacuum Trap System for Aspiration and Vacuum Protection, Foxx Life Sciences
Vacuum Traps
Replace outdated glass vacuum traps with a shatterproof, pre-assembled system.
  • Comes ready-to-use, no assembly required
  • Shatterproof bottles offer improved safety
  • VersaCap eliminates tube tangling
  • Easy to empty without detaching tubing
  • Inline hydrophobic filter included
  • Optional ultra-stable secondary container
  • Optional overflow bottle for CDC compliance
The Vactrap™ 2 vacuum trap system adheres to the strictest CDC recommendations for laboratory safety in vacuum applications.
Ideal for protecting vacuum lines and users, Vactrap™ safely containsMore Product Information