Anti-Camelid VHH Rabbit Monoclonal Antibody [clone: 96A3F5] (iFluor 488)
Supplier: Genscript
A01862-200
76197-320EA 408.57 CAD
76197-320
Anti-Camelid VHH Rabbit Monoclonal Antibody [clone: 96A3F5] (iFluor 488)
Antibodies
GenScript MonoRab™ Rabbit Anti-Camelid VHH Antibody [iFluor488], mAb is specific for Camelid VHH Antibody (Alpaca, Llama and Camel). The monoclonal antibody has no cross-reactivity with mouse, rat, rabbit, chicken, goat or human immunoglobulins.
  • Anti-VHH Antibodies
  • MonoRab™ technology guarantees high affinity: Kd ~10 to 11 M
  • No cross reactivity with other species
  • Specific to camelid IgG2 and IgG3
  • Recognize conformational epitopes of the VHH domain
  • Recognize both camelid and humanized VHHs
GenScript has developed a series of rabbit monoclonal (MonoRab)More Product Information