Top Dog 6 Mil Sleeves, 18" Length, Remco Products
Supplier: Remco Products
84012 84013 84014 84015 84016
76196-526CS 495.14 CAD
76196-526 76196-528 76196-530 76196-532 76196-534
Top Dog 6 Mil Sleeves, 18" Length, Remco Products
Workwear
Reusable and washable, the one-piece construction increases the sleeve's hygiene and durability.
  • Resistance to chemical and acid sanitation products
  • Non-porous surface will not host bacteria or mold
  • Washer and dryer safe
  • Will not crack or stiffen in low temperatures or with repeated cleaning
  • Impervious to animal fats and degreasers
  • Completely latex-free for sensitive users
  • Easily repairable
These sleeves are made of 6 mil pure polyurethane. They provide coverage from above the elbow to the wrist, with elastic at both ends to keep them in place. FiveMore Product Information