TC Bozemann Wertheim Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-8093
76192-682EA 2432.48 CAD
76192-682
TC Bozemann Wertheim Needle Holder, OR Grade, Sklar®
Forceps
The Sklar® Tungsten Carbide Bozeman needle holder is commonly used for suturing in gynecological procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The shanks of this instrument are slightly curved and therefore provide greater visibility when used in the uterine cavity. Cross-serrated tips and a ratchet mechanism securely hold needles in place. TCMore Product Information