Econo™ Tuttle Forceps, Floor Grade, Sklar
Supplier: Sklar
21-617
76192-612BX 362.62 CAD
76192-612
Econo™ Tuttle Forceps, Floor Grade, Sklar
Forceps
Econo™ Tuttle Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Econo™ Tuttle Forceps have long shafts which make them useful for working in areas such as the intestines. These forceps may also be known as SingleyMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable