Supercut Sklarhone™ Solz 'Gold Tip' Scissors, OR Grade, Sklar
Supplier: Sklar
15-9301 15-9302 15-9303
76192-030EA 1525.28 CAD
76192-030 76192-032 76192-034
Supercut Sklarhone™ Solz 'Gold Tip' Scissors, OR Grade, Sklar
Scissors
Sklar's® Solz gold tip scissors, also know as Kaye or Sims, are a dissecting scissor used in plastic surgery procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The design of the shank facilitates ideal atraumatic seperation of the tissue layers. Supercut Sklarhone™ scissors combine one knife-like razor edge and one micro-serrated scissor edge. TheseMore Product Information