Esteem® Stretchy Nitrile Examination Gloves, Cardinal
Supplier: Cardinal Health
8815NB 8854NXSB 8855NSB 8857NLB N8854XPB N8850XPB N8853XPB N8852XPB N8851XPB
PHN8852XPCS 399.8 CAD
PHN8852XP PH8815NB PH8816NB PH8817NB PH8819NB PH8854NXSB PH8855NSB PH8856NMB PH8857NLB PH8858NXLB PH8859NXXB 76183-708 76183-710 76183-730 76183-732 76183-734
Esteem® Stretchy Nitrile Examination Gloves, Cardinal
Gloves
Powder-free nitrile gloves are ideal for applications requiring maximum tactility and sensitivity. Extra-stretchy nitrile formulation ensures a more comfortable fit than other nitrile gloves and conforms to hands quickly to facilitate a wide range of motion, especially around the thumb area. Gloves are puncture-resistant and have textured fingertips for a firm, no-slip grip. Beaded cuff resists tearing and minimizes cuff rolldown. Ambidextrous.
Textured gloves have a smooth surface and textured fingers.XP gloves have the same AQL as surgical gloves to ensure barrier protection.