Cole-Parmer® Grinding Vial Sets
Supplier: SPEX
3114 5004I 5004W 6751 6751C20 6751C4 6751E 6757 6759 6761 6761E 6761P 6771 6771E 6771P 6781E 6801C20 6801C4 6801E 6801P 6802 6871 6871E 6871P 6881 6881C20 6881C4 6881E 6881P 6883 6883E 6885P 8001 8004SS 8005 8008 8009 6751P 6753C 6781O 6781S 6801 8004 2240-PC 2240-PEF 2250 3117 3120 3127 8007 5004 5004C 5004E 6757C3 6757E 6757P 6883P 6885 6885E 8014 2254 5005 5006 6803S 8001LC 8020
76181-856EA 257.65 CAD
76181-856 76181-938 76181-940 76181-950 76181-952 76181-954 76181-956 76181-960 76181-962 76181-964 76181-966 76181-968 76181-970 76181-972 76181-974 76181-980 76181-986 76181-988 76181-990 76181-992 76181-994 76181-998 76182-000 76182-002 76182-016 76182-018 76182-020 76182-022 76182-024 76182-026 76182-028 76182-030 76182-068 76182-102 76182-104 76182-108 76182-112 76182-222 76182-224 76182-230 76182-232 76182-236 76182-250 76182-296 76182-298 76182-304 76182-352 76182-360 76182-362 76182-390 76182-416 76182-420 76182-422 76182-424 76182-426 76182-428 76182-448 76182-452 76182-454 76182-466 76182-512 76533-082 76533-114 76533-116 76533-138 76533-140 76533-144
Cole-Parmer® Grinding Vial Sets
Homogenizers Benchtop Homogenizers
Ideal for pulverizing the toughest rocks, mineral, cement, and ceramics.
  • Optimize performance of your Cole-Parmer® grinders with a wide range of vial sets suitable for your application
  • Keep spare parts to ensure there is no down time
Choose grinding vial sets and grinder balls suitable for use with the Spex® SamplePrep 1200 GenoLyte® compact homogenizer, 5120 Mixer/Mill®, 8000M Mixer/Mill®, 8000D Dual Mixer/Mill®, and other grinding equipment.
Order Now
SPECIFICATIONS
Environmentally Preferable