Cole-Parmer® X-Press® XP-400 Hydraulic Laboratory Pellet Press
Supplier: SPEX
3613 3613ST 3614 3614C 3614ST 3614W 3616 3616C 3616ST 3616W 3636-230 3623 3623C 3623ST 3623W 3625 3636-115
76181-890EA 2258.82 CAD
76181-890 76181-892 76181-894 76181-896 76181-898 76181-900 76181-906 76181-908 76181-910 76181-912 76181-930 76182-386 76182-388 76182-398 76182-400 76182-402 76182-406
Cole-Parmer® X-Press® XP-400 Hydraulic Laboratory Pellet Press
Presses
The Cole-Parmer® XP-400 X-Press® hydraulic laboratory pellet press is an automatic and fully programmable pellet press designed for repetitive pressing of sample pellets for XRF, IR, and other analytical techniques. This press can also be operated manually as a simple motorized press whose functions are under direct user control.
  • Ideal for accurate and repetitive pressing of sample pellets for XRF and other analytical techniques
  • Automated, programmable laboratory press allows for samples to be pressed within minutes
  • Ensure safe operation with safety
More Product Information