Cole-Parmer® HG-600 Geno/Grinder® 2010 Tissue Homogenizers and Cell Lysers
Supplier: SPEX
2010-115 2010-230
76181-808EA 44424.97 CAD
76181-808 76181-810
Cole-Parmer® HG-600 Geno/Grinder® 2010 Tissue Homogenizers and Cell Lysers
Homogenizers Tissue Grinders
The Geno/Grinder® is an automated high-throughput plant and animal tissue homogenizer and cell lyser.
  • Fast and efficient extraction of nucleic acids, proteins and other molecules
  • Double-locking lid provides safe operation
  • Sound insulation and suspension system ensure quiet, vibration-free operation
  • Fully adjustable clamp allows simple front loading of vials or microplates
  • Interior LED lighting and large window improve sample visibility
  • Fully programmable control screen tilts for optimal viewing angle
The HG-600 Geno/Grinder® 2010 high-throughputMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable