VWR® Rotors for VWR® Laboratory Centrifuges
Supplier: VWR International
76180-952 76180-956 76181-192 76181-194 76181-196 76181-198 76181-200 76181-202 76181-206 C3303-0450H
76180-952PK 460.73 CAD
76180-952 76180-956 76181-192 76181-194 76181-196 76181-198 76181-200 76181-202 76181-206 76457-572
VWR® Rotors for VWR® Laboratory Centrifuges
Centrifuges Rotors
Swing out or angled rotors for separation.
  • Rotors for horizontal or angled separation
  • Accepts smaller tubes with the use of adapters
Swing out rotors: The 4×50 ml horizontal rotor and the 6×15 ml horizontal rotor of the VWR® Laboratory Centrifuge are the perfect accessories for horizontal separation of 50 ml tubes and 15 ml tubes respectively. These rotors will also accept smaller tubes with the use of optional adapters.
Angled rotors: Rotors for angled separation include 18×10 ml, 24×10 ml, 12×15 ml, 24×15 ml and 6×50 ml. These rotors will alsoMore Product Information