PIPETMAN® Diamond Tips, Sterilized Filter Tipack™ Racks, Gilson®
Supplier: GILSON, INC.
F171203 F171103 F171303 F171403 F171503 F171603 F171703 F171803
76178-212PK 362.93 CAD
76178-212 76178-214 76178-216 76178-218 76178-220 76178-222 76178-224 76178-226
PIPETMAN® Diamond Tips, Sterilized Filter Tipack™ Racks, Gilson®
Pipette Tips
PIPETMAN® DIAMOND Sterilized Filter Tips are designed to fit your PIPETMAN perfectly and play a critical role in preventing aerosol-born contamination. They deliver accurate and precise pipetting when used in conjunction with Gilson pipettes.
  • Accurate and precise
  • Reliable
  • Convenient packaging
Using PIPETMAN DIAMOND Tips with your PIPETMAN ensures maximum accuracy and precision and delivers consistent results. The perfect air-tight seal prevents leaks. The tips are made of pure polypropylene to deliver the correct volume and ensure complete dispensing.More Product Information