PIPETMAN® Diamond Tips, Autoclavable, in Tipack™ Racks, Gilson®
Supplier: GILSON, INC.
F171100 F171200 F171300 F171400 F171500 F171600 F161370 F161230 F171101 F171201 F171301 F171401 F171501 F171601
76178-184PK 167.26 CAD
76178-184 76178-186 76178-188 76178-190 76178-192 76178-194 76178-196 76178-198 76178-200 76178-202 76178-204 76178-206 76178-208 76178-210
PIPETMAN® Diamond Tips, Autoclavable, in Tipack™ Racks, Gilson®
Pipette Tips
PIPETMAN® DIAMOND Tips are designed to fit your PIPETMAN perfectly. They deliver accurate and precise pipetting when used in conjunction with Gilson pipettes.
  • Accurate and precise
  • Reliable
  • Convenient packaging
Using PIPETMAN DIAMOND Tips with your PIPETMAN ensures maximum accuracy and precision and delivers consistent results. The perfect air-tight seal prevents leaks. The tips are made of pure polypropylene to deliver the correct volume and ensure complete dispensing. Volume graduations make it easy to visually check the volume aspirated.
Order Now
SPECIFICATIONS
Environmentally Preferable