PIPETMAN® Diamond Tips EcoPack™, Gilson®
Supplier: GILSON, INC.
F161630 F161450 F161930 F161730 F161670 F161110
76178-170PK 939.61 CAD
76178-170 76178-172 76178-174 76178-176 76178-178 76178-574
PIPETMAN® Diamond Tips EcoPack™, Gilson®
Pipette Tips
PIPETMAN® DIAMOND Tips are designed to fit your PIPETMAN perfectly. They deliver accurate and precise pipetting when used in conjunction with Gilson pipettes.
  • Accurate and precise: Using PIPETMAN DIAMOND Tips with your PIPETMAN ensures maximum accuracy and precision and delivers consistent results
  • Reliable: Our tips are manufactured in a clean room environment ensuring they are free of detectable contaminants that can affect your experiments
  • Economical: Ordering in bulk and simply reloading the tips into empty racks saves you money
PIPETMAN® DIAMONDMore Product Information