PIPETMAN® Diamond EasyPack™ Tips, Gilson®
Supplier: GILSON, INC.
F161930/1 F161730/1 F161630/1 F161450/1 F161670/1
76178-150PK 106.65 CAD
76178-150 76178-154 76178-576 76178-580 76180-022
PIPETMAN® Diamond EasyPack™ Tips, Gilson®
Pipette Tips
PIPETMAN® DIAMOND Tips are designed to fit your PIPETMAN perfectly and deliver accurate and precise pipetting when used in conjunction with Gilson pipettes.
  • Accurate and precise
  • Reliable
  • Convenient use and storage
  • Minimal waste
  • Fully autoclavable
Using PIPETMAN DIAMOND Tips with your PIPETMAN ensures maximum accuracy and precision and delivers consistent results. The perfect air-tight seal prevents leaks. The tips are made of pure polypropylene to deliver the correct volume and ensure complete dispensing. Volume graduations make it easy to visuallyMore Product Information