PIPETMAN® Single Channel Pipettor, Gilson®
Supplier: GILSON, INC.
F144054M F144055M F144056M F144057M F144058M F144059M F144066 F144067
76178-030EA 983.74 CAD
76178-030 76178-032 76178-034 76178-036 76178-038 76178-040 76178-042 76178-044
PIPETMAN® Single Channel Pipettor, Gilson®
Pipettes Micropipettes
PIPETMAN®, recognized as the pipetting standard for its reliability and proven durability, is a fully adjustable, air-displacement pipette that uses disposable tips. PIPETMAN combines legendary accuracy, precision, and durability with easy pipetting and handling.
  • Legendary performance
  • Easy pipetting
  • Built-to-last and convenient
  • Simple to maintain
PIPETMAN pipettes come in a large range of volumes with reduced pipetting forces for hours of comfortable pipetting. Eight single channel models cover a volume range from 0.2 µl to 10 ml and eightMore Product Information