VWR® Covers
Supplier: VWR International
76176-376 76176-378 76176-390 76176-392 30107 76176-396 30110 76176-400 30116 76176-404 30200S 30200 76176-410 76176-412 76176-414 76176-416 30205 76176-420 30207 30208 76176-426 76176-428 76176-430 76176-432 30210 30212 30216 76176-440 76176-442 30300 76176-446 76176-448 76176-450 76176-452 76176-454 76176-456 30307 76176-460 76176-462 76176-464 76176-466 30318 35100S 35100 35203 35204 35205 35206 35207 35208 35210 35212 35216 35218 35300S 35300 35301 76176-500 76176-502 76176-504 76176-506 35306 35307 35308 76176-514 76176-516 35316 35318 310100SB 310100B 76176-526 76176-528 310107B 310108B 35110B 35112B 35116B 35118B 310200SB 310200B 310201B 310202B 310203B 310204B 310205B 310206B 310207B 310208B 35210B 35212B 35216B 35218B 310300SB 76176-572 76176-574 76176-576 76176-578 310304B 76176-582 310306B 310307B 310308B 35310B 35312B 35316B 35318B 76176-598 76176-600 76176-602 76176-604 35105 35106 35107 35108 76176-614 35112 35116 35118 35200S 35200 35201 35202 310103B 76176-632 310105B 310106B 76205-172 76205-174 76205-176 76205-178 76205-180 76205-182 76205-184
76176-376CS 300.9 CAD
76176-376 76176-378 76176-390 76176-392 76176-394 76176-396 76176-398 76176-400 76176-402 76176-404 76176-406 76176-408 76176-410 76176-412 76176-414 76176-416 76176-418 76176-420 76176-422 76176-424 76176-426 76176-428 76176-430 76176-432 76176-434 76176-436 76176-438 76176-440 76176-442 76176-444 76176-446 76176-448 76176-450 76176-452 76176-454 76176-456 76176-458 76176-460 76176-462 76176-464 76176-466 76176-468 76176-470 76176-472 76176-474 76176-476 76176-478 76176-480 76176-482 76176-484 76176-486 76176-488 76176-490 76176-492 76176-494 76176-496 76176-498 76176-500 76176-502 76176-504 76176-506 76176-508 76176-510 76176-512 76176-514 76176-516 76176-518 76176-520 76176-522 76176-524 76176-526 76176-528 76176-530 76176-532 76176-534 76176-536 76176-538 76176-540 76176-542 76176-544 76176-546 76176-548 76176-550 76176-552 76176-554 76176-556 76176-558 76176-560 76176-562 76176-564 76176-566 76176-568 76176-570 76176-572 76176-574 76176-576 76176-578 76176-580 76176-582 76176-584 76176-586 76176-588 76176-590 76176-592 76176-594 76176-596 76176-598 76176-600 76176-602 76176-604 76176-606 76176-608 76176-610 76176-612 76176-614 76176-616 76176-618 76176-620 76176-622 76176-624 76176-626 76176-628 76176-630 76176-632 76176-634 76176-636 76205-172 76205-174 76205-176 76205-178 76205-180 76205-182 76205-184
VWR® Covers
Covers
VWR® Covers provide a convenient means to cover and protect bowls, flanges, flasks, beakers, containers, media bottles, and hoses
  • Breathable
  • High-quality construction
  • Form fitting
  • Variety of sizes
  • Made in the USA
Covers utilize top materials such SMS, Alliance®, and Tyvek®, which offer a variety of beneficial characteristics.
Ordering information: If the size and material combination required is not listed, simply contact your Avantor Controlled Environment Specialist to assist.