TORBAL FSC Tool and Wrench Torque Gauges, Scientific Industries
Supplier: Scientific Industries
FSC2 FSC5 FSC10
76175-354EA 2756.59 CAD
76175-354 76175-356 76175-358
TORBAL FSC Tool and Wrench Torque Gauges, Scientific Industries
Dynamometers
The FSC is an advanced precision torque tester ideal for accurate verification or adjustment of virtually any manual or electric wrench, and screwdriver. Easily adaptable to almost any working environment and processes.
  • Built-in wireless
  • 1000 Hz data capture
  • MicroSD card
  • USB and RS232 interface
  • Analog I/O modes
  • Rugged design
  • Stats and analytics
The FSC TORQUE ranges in capacity from 17 lbF.in (2 Nm) with a resolution of 0.01 lbF.in (0.001 Nm) to 100 lbF.in (10 Nm) with a resolution of 0.1 lbF.in (0.01 Nm)
The Pass / Fail threshold settings makesMore Product Information