AlphaTec® 58-735 Medium-Duty Chemical and Cut Protection Gloves, Ansell
Supplier: Ansell Healthcare
58735070 58735080 58735090 58735100 58735110
76170-082CS 3437.48 CAD
76170-082 76170-084 76170-268 76170-270 76170-272
AlphaTec® 58-735 Medium-Duty Chemical and Cut Protection Gloves, Ansell
Gloves
Advanced chemical protection with a great level of cut defense.
  • INTERCEPT™ cut resistant technology provides protection against lacerations
  • Optimized fit integrates the cut liner into a nitrile shell for better dexterity and tactility versus double gloving
  • Excellent dry and wet grip enhance dexterity and reduce hand fatigue
  • High-visibility cut liner acts as an indicator for when glove is cut as chemical protection is compromised
  • Cuff can be folded to act as a gutter to prevent chemicals running down the arm
  • Liner material: Ansell INTERCEPT™ Technology
Order Now
SPECIFICATIONS
Environmentally Preferable
Ambidextrous/Hand-specific Hand-specific
Disposable / reusable Reusable
Latex/Latex-free Latex
Powdered/Powder-free Powder-free
Gauge 18
Anti-static +