Full Size Wireless Nylon Tube Racks, Globe Scientific
Supplier: Globe Scientific
456540R 456540W 456540Y 456640B 456640G 456640M 456640N 456640R 456640W 456440B 456440G 456440M 456640Y 456740B 456740G 456740M 456740N 456740R 456740W 456740Y 456840B 456840G 456840M 456540G 456540M 456540N 456440N 456440R 456440W 456440Y 456540B 456840N 456840R 456840W 456840Y
76169-926EA 47.57 CAD
76169-926 76169-928 76169-930 76169-932 76169-934 76169-936 76169-938 76169-940 76169-942 76169-944 76169-946 76169-948 76170-008 76170-010 76170-012 76170-014 76170-016 76170-018 76170-020 76170-022 76170-024 76170-026 76170-028 76170-086 76170-088 76170-090 76170-092 76170-094 76170-096 76170-098 76170-100 76170-196 76170-198 76170-200 76170-202
Full Size Wireless Nylon Tube Racks, Globe Scientific
Racks Tube Racks
These fully autoclavable racks are molded from reinforced nylon for strength and durability.
  • Single piece, three tier grid design for secure and stable racking of tubes
  • Good temperature and chemical resistance to withstand a variety of lab procedures
  • Large, flat endplates for labeling and easy sample identification
  • Dense nylon material will not float in water bath
  • 15 ml conicals fit in 20 mm racks and 50 ml conicals fit in 30 mm racks
  • Seven colorful options to keep samples and benchtop organized
With no coatings to peel or chip, the wireless tubeMore Product Information