Microscope Slide Storage Box with Removable Tray, Globe Scientific
Supplier: Globe Scientific
513250W 513250P 513250Y 513250B 513250G 513252W 513252P 513252Y 513252B 513252G
76169-976CS 189.36 CAD
76169-976 76169-978 76169-980 76169-982 76169-984 76169-986 76169-988 76169-990 76169-992 76169-994
Microscope Slide Storage Box with Removable Tray, Globe Scientific
Microscope Slide Boxes
The rugged and durable microscope slide storage boxes feature a hinged lid for protection while providing easy viewing and access to your microscope slides.
  • Transparent lid provides clear view of contents without opening
  • Removable draining tray holds 100 slides in pre-numbered slots
  • Tray provides space between each slide so slides can be tilted forward or back for easy identification
The trays can accommodate up to 200 standard sized microscope slides (2 I each pre-numbered slot) on end for space savings.
Ordering information: Removable drainingMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable