Cool Offs, Allegro
Supplier: Allegro
8405-50 8405-51 8405-53 8405-54 8405-55 8405-59
76169-212EA 4.38 CAD
76169-212 76169-214 76169-216 76169-218 76169-220 76169-222
Cool Offs, Allegro
Workwear
Cooling products are designed to protect and provide comfort to workers who must function in environments with excessive heat.
  • Lightweight and comfortable
  • 38" length (longest on the market)
  • 100% cotton
Keep cool all day long with these dual-use Cool Offs
To use, simply soak the garment in cold water for 10 to 15 minutes. The cooling crystals go to work to keep wearers cool for days.
Quilted into four sections for an even distribution of hydration material, Cool Offs can be worn around the neck or tied as a headband. A unique button hole closureMore Product Information