BACTRON Anaerobic Chamber, 900 Plate Capacity, Sheldon
Supplier: Sheldon Manufacturing
BAA90022 BAA90022-E
76097-900EA 81043.56 CAD
76097-900 76097-914
BACTRON Anaerobic Chamber, 900 Plate Capacity, Sheldon
Anaerobic Systems
BACTRON chambers are total anaerobic process workstations. Samples can be prepared, cultured, and inspected in the oxygen-free chamber and standard incubator. Unlike anaerobic jars and bags, which unavoidably expose samples to oxygen, BACTRON units provide complete anaerobic environments for all your applications.
  • Ergonomic arm ports provide a comfortable work space
  • Integraded vacuum pump for quieter operation
  • 60% faster airlock cycle with 3,4 or 5 auto cycle iterations
  • Auto-sleeve cycle with integrated HEPA filter, 40% faster sleeve cycle
  • Low gas supply
More Product Information