VWR® Eco Racks
Supplier: VWR International
76051-512 76051-514 76051-516 76051-518 76051-520 76051-522 76051-524 76051-526 76051-528
76051-512EA 112.86 CAD
76051-512 76051-514 76051-516 76051-518 76051-520 76051-522 76051-524 76051-526 76051-528
VWR® Eco Racks
Racks Cryogenic Box Racks
Eco rack for 2" boxes.
  • Stainless steel
  • Double welds
  • Smoothed edges
  • Lifetime warranty
These freezer racks are made to the highest levels of quality with pharmaceutical grade stainless steel, double welds, smoothed edges and an unconditional lifetime warranty.