SW® MegaMan® MM-11BK Black 10.1 mil Sustainable Sweat-Absorbing Nitrile Exam Gloves
Supplier: SW Safety Solutions
N260881 N260882 N260883 N260884 N260885 N260886 N260887
76051-152BX 39.68 CAD
76051-152 76051-154 76051-210 76051-212 76051-214 76051-216 76051-218
SW® MegaMan® MM-11BK Black 10.1 mil Sustainable Sweat-Absorbing Nitrile Exam Gloves
Gloves
These gloves combine sweat management and unprecedented strength in a fully-textured glove that delivers the ultimate in comfort and performance.
  • DriTek® comfort technology with absorbent lining that wicks away moisture for added extended use
  • EcoTek® sustainable technology
  • ANSI/EN level 3 abrasion resistance for durability and extended wear
  • Fully textured surface for excellent grip
  • Optimal thickness for durability and tactile sensitivity
  • 1.5 AQL exceeds laboratory standards
  • Up to 3× longer lasting than standard nitrile gloves
  • SHA dermatologically approved
More Product Information