SW® PowerForm® PF-95TL Teal 5.8 mil Sustainable Nitrile Exam Gloves
Supplier: SW Safety Solutions
N200362 N200363 N200364 N200365 N200366 N200361
76051-116BX 26.05 CAD
76051-116 76051-118 76051-120 76051-122 76051-124 76554-054
SW® PowerForm® PF-95TL Teal 5.8 mil Sustainable Nitrile Exam Gloves
Gloves
PowerForm® gloves are versatile and sustainable 5 mil single-use glove combines maximum comfort and control with high-quality construction and high-performance protection.
  • EcoTek® sustainable technology
  • Fully textured for excellent wet and dry grip
  • Touch-screen compatible
  • Silicone-free for critical environments
  • Optimal thickness for durability and tactile sensitivity
  • Tear-resistant beaded cuff eases donning
  • 1.5 AQL exceeds laboratory standards
  • Excellent price/performance
EcoTek® gloves tested to ASTM D5526-12 standard test method for determiningMore Product Information