PTFE Valves, Two-Way and Three-Way, Burkle
Supplier: BURKLE INC
8610-0006 8610-0010 8610-0012 8610-0015 8610-0001 8610-0003
76049-254EA 307.01 CAD
76049-254 76049-256 76049-258 76049-260 76049-400 76049-402
PTFE Valves, Two-Way and Three-Way, Burkle
Valves Vacuum Valves
Two-way/Three-way valves are made of solid PTFE with hose nozzles
  • Resistant to compression; 1 bar (14.50 psi) maximum
  • Vacuum suitable up to 10-1 mmHg
  • Autoclavable
They are resistant to chemicals, and offer very easy connection due to excellent anti-friction properties. Blue handles are made of polypropylene.