Protector® ClassMate™ Laboratory Hoods, Labconco®
Supplier: Labconco
160404121 160405100 160405101 160405102 160405120 160504102 160504120 160504121 160505100 160505101 160505102 160505120 160505121 160604100 160604101 160604102 160604120 160604121 160605100 160605101 160605102 160405121 160504100 160504101 160404100 160404101 160404102 160404120 160605120 160605121
76045-616EA 20026.5 CAD
76045-616 76045-618 76045-620 76045-622 76045-624 76045-626 76045-628 76045-630 76045-632 76045-634 76045-636 76045-638 76045-640 76045-642 76045-644 76045-646 76045-648 76045-650 76045-652 76045-654 76045-656 76045-906 76045-908 76045-910 76045-912 76045-914 76045-916 76045-918 76045-920 76045-922
Protector® ClassMate™ Laboratory Hoods, Labconco®
Fume Hoods and Enclosures Chemical Fume Hoods
ClassMate™ laboratory hoods are designed to meet the needs of instructional laboratories.
  • High-performance, bypass airflow design
Clear tempered safety glass provides excellent visibility even when hoods are placed back to back
The hood frame, spill trough, and standard hood air foil are constructed of glacier white powder-coated steel and aluminum. The low-profile spill trough is 3.2 cm (11/4”) deep and requires no cabinet overhang. All hoods include pre-wired LED lighting, ADA-compliant light and blower switches, removable interior cover platesMore Product Information