VWR® Cell Culture Chamber, Sterile
Supplier: VWR International
76045-394 76045-402 76045-404 76045-410
76045-394CS 737.74 CAD
76045-394 76045-402 76045-404 76045-410
VWR® Cell Culture Chamber, Sterile
Bioreactors
This sterile cell culture chamber, which is available with 1, 2, 5, or 10 layers, produces large-scale cell culture results in just one cycle.
  • Sterilized by irradiation
  • Non-pyrogenic and Dnase-/RNase-free
The device is assembled by hydrophobic membranes with an aperture of 0.22 µm to avoid potential contamination in gas exchange. The device does not absorb cytokines and growth factors, and the conventional trypsin digestion method is applied in cell harvesting. Two 26 mm diameter filling ports allow direct access to the chamber bottom, providingMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M