Lab Vortex Mixers with Touch Function

Supplier: Heathrow Scientific
HS120318 HS120209
76027-582EA 694.15 CAD
76027-582 76027-828
Lab Vortex Mixers with Touch Function
Shakers Vortex Mixers
This unique and innovative designed Vortex Mixer is a variable speed vortexing unit with multi-purpose and maxium convenience. Why take time to search for separate mixing heads for various sized tubes when this vortex mixer provides one mixing head that will accomodate tubes ranging from 0.2 mL - 50 mL and a standard assay plate.

  • Variable speed rotary dial provides pre-set speed control (1000, 2000, 3000 rpm) during vortex mixing
  • Stable elastomeric device feet, provide vibration dampening and prevent movement during operation
  • Innovative head can be removed
More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Speed range 1000 - 3000 rpm
Weight 8.6 lbs.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR